Learn More
Med Vet International Csr Sterilization Wrap, 18"X18", 50/Pack
Catalog No. 502830980
Encompass_Preferred
Our disposable autoclavable wrappers are superior to muslin and are virtually memory free. The natural construction of these disposable wrappers enables the user to wrap continuously without fear of skin sensitization. These products breathe easily and are also highly liquid repellent. Can be used as surgical drape.
Catalog No. 50-283-0980 Supplier Med Vet International Supplier No. 141850
Our disposable autoclavable wrappers are superior to muslin and are virtually memory free. The natural construction of these disposable wrappers enables the user to wrap continuously without fear of skin sensitization. These products breathe easily and are also highly liquid repellent. Can be used as surgical drape.