Learn More
Med Vet International CSR Wrap, Single-Ply, 20-in x 20-in 1000/Case
Catalog No. 503044422
Encompass_Preferred
Suitable for light trays, small linen packs, small medical devices and small instruments Delivers high-level protection against fluids and particulates for exceptional strength and improved efficiency Reliable low linting fabric to help maintain the integrity of the sterile packs and create an ideal surface for sterilization tape Durable fabric resists tears, yet still soft for easy draping
Catalog No. 50-304-4422 Supplier Med Vet International Supplier No. 1420
Suitable for light trays, small linen packs, small medical devices and small instruments Delivers high-level protection against fluids and particulates for exceptional strength and improved efficiency Reliable low linting fabric to help maintain the integrity of the sterile packs and create an ideal surface for sterilization tape Durable fabric resists tears, yet still soft for easy draping