Learn More
Med Vet International Csr Sterilization Wrap, 45"X45", 50/Pack
Catalog No. 502831162
Encompass_Preferred
Our disposable autoclavable wrappers are superior to muslin and are virtually memory free. The natural construction of these disposable wrappers enables the user to wrap continuously without fear of skin sensitization. These products breathe easily and are also highly liquid repellent. Can be used as surgical drape.
Catalog No. 50-283-1162 Supplier Med Vet International Supplier No. 144550
Our disposable autoclavable wrappers are superior to muslin and are virtually memory free. The natural construction of these disposable wrappers enables the user to wrap continuously without fear of skin sensitization. These products breathe easily and are also highly liquid repellent. Can be used as surgical drape.