Learn More
Med Vet International Dynarex Sensi Wrap, Self-Adherent, Latex Free, 6" X 5 Yards, Assorted, 12/Case
Catalog No. 502831974
Encompass_Preferred
Ideal for reducing edema and for holding splints, gel sheeting or elastomer molds in place, available in individual colors. A lightweight compression bandage ideal for treatment of a variety of injuries including sprains and relief bandaging of joints and other difficult-to-wrap areas No clips or fasteners needed; adhesive wrap sticks to itself, not to other materials or skin Will not slip - layers "bond" together for the strongest holding power Easily torn without scissors Individually poly bagged Each assorted pack contains the following colors: red, green, purple, dark blue, pink and light blue. Not made with natural rubber latex
Catalog No. 50-283-1974 Supplier Med Vet International Supplier No. 3219
Ideal for reducing edema and for holding splints, gel sheeting or elastomer molds in place, available in individual colors. A lightweight compression bandage ideal for treatment of a variety of injuries including sprains and relief bandaging of joints and other difficult-to-wrap areas No clips or fasteners needed; adhesive wrap sticks to itself, not to other materials or skin Will not slip - layers "bond" together for the strongest holding power Easily torn without scissors Individually poly bagged Each assorted pack contains the following colors: red, green, purple, dark blue, pink and light blue. Not made with natural rubber latex