Learn More
Med Vet International Quick Check* H500 Sterilization Wrap White / Blue 24 X 24 inch Dual Layer Sms Polypropylene Steam / Eo Gas / Hydrogen Peroxide, 34162, 120/Case
Catalog No. 502829488
Encompass_Preferred
Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Use with heavy instrument sets, retractors, sets where frequent handling occurs Made from recyclable number 5 polypropylene fabric 2 x 2 feet There are 120 blue-and-white sterilization wraps in a case
Catalog No. 50-282-9488 Supplier Med Vet International Supplier No. 809418CS
 Restricted Item
Add to Cart
Add to Cart
Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Use with heavy instrument sets, retractors, sets where frequent handling occurs Made from recyclable number 5 polypropylene fabric 2 x 2 feet There are 120 blue-and-white sterilization wraps in a case