Learn More
Med Vet International Quick Check* H400 Sterilization Wrap White / Blue 24 X 24 inch Dual Layer Sms Polypropylene Steam / Eo Gas / Hydrogen Peroxide, 34183, 60/Pack
Catalog No. 502830804
Encompass_Preferred
Suggested uses based on wrap grade: Most basin/instrument sets, procedure trays and drills Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Use with most basin or instrument sets, procedure trays, drills Made from recyclable number 5 polypropylene fabric 2 x 2 feet
Catalog No. 50-283-0804 Supplier Med Vet International Supplier No. 810872PK
 Restricted Item
Add to Cart
Add to Cart
Suggested uses based on wrap grade: Most basin/instrument sets, procedure trays and drills Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Use with most basin or instrument sets, procedure trays, drills Made from recyclable number 5 polypropylene fabric 2 x 2 feet