Learn More
Med Vet International Quick Check* H600 Sterilization Wrap White / Blue 36 X 36 inch Dual Layer Sms Polypropylene Steam / Eo Gas / Hydrogen Peroxide, 34151, 36/Pack
Catalog No. 502829623
Encompass_Preferred
Comes with white and blue layers that are bonded together – inspect trays more easily and quickly than ever Superior microbial barrier protection with patented Powerguard technology Made from recyclable number 5 polypropylene fabric Use for extra heavy instrument sets, loaner trays
Catalog No. 50-282-9623 Supplier Med Vet International Supplier No. 810875PK
Comes with white and blue layers that are bonded together – inspect trays more easily and quickly than ever Superior microbial barrier protection with patented Powerguard technology Made from recyclable number 5 polypropylene fabric Use for extra heavy instrument sets, loaner trays