Learn More
Med Vet International Quick Check* H500 Sterilization Wrap White / Blue 36 X 36 inch Dual Layer Sms Polypropylene Steam / Eo Gas / Hydrogen Peroxide, 34199, 72/Case
Catalog No. 502830932
Encompass_Preferred
Suggested uses based on wrap grade: Heavy instrument sets, retractors, sets where frequent handling occurs Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Use with most basin or instrument sets, procedure trays, drills Made from recyclable number 5 polypropylene fabric 1 x 1 yard
Catalog No. 50-283-0932 Supplier Med Vet International Supplier No. 810873CS
Suggested uses based on wrap grade: Heavy instrument sets, retractors, sets where frequent handling occurs Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Use with most basin or instrument sets, procedure trays, drills Made from recyclable number 5 polypropylene fabric 1 x 1 yard