Learn More
Med Vet International Quick Check* H600 Sterilization Wrap White / Blue 45 X 45 inch Dual Layer Sms Polypropylene Steam / Eo Gas / Hydrogen Peroxide, 34164, 48/Case
Catalog No. 502830807
Encompass_Preferred
Suggested uses based on wrap grade: Extra heavy instrument sets, loaner trays Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Use with most basin or instrument sets, procedure trays, drills Made from recyclable number 5 polypropylene fabric 45 x 45 inches
Catalog No. 50-283-0807 Supplier Med Vet International Supplier No. 810876CS
Suggested uses based on wrap grade: Extra heavy instrument sets, loaner trays Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Use with most basin or instrument sets, procedure trays, drills Made from recyclable number 5 polypropylene fabric 45 x 45 inches