Learn More
Med Vet International Quick Check* H500 Sterilization Wrap White / Blue 45 X 45 inch Dual Layer Sms Polypropylene Steam / Eo Gas / Hydrogen Peroxide, 34198, 48/Case
Catalog No. 502830806
Encompass_Preferred
Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Use with heavy instrument sets, retractors, sets where frequent handling occurs Made from recyclable number 5 polypropylene fabric
Catalog No. 50-283-0806 Supplier Med Vet International Supplier No. 810874CS
Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Use with heavy instrument sets, retractors, sets where frequent handling occurs Made from recyclable number 5 polypropylene fabric