Learn More
Med Vet International Quick Check* H600 Sterilization Wrap White / Blue 48 X 48 inch Dual Layer Sms Polypropylene Steam / Eo Gas / Hydrogen Peroxide, 34147, 48/Case
Catalog No. 502831999
Encompass_Preferred
Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Made from recyclable number 5 polypropylene fabric Use for extra heavy instrument sets, loaner trays 4 x 4 feet
Catalog No. 50-283-1999 Supplier Med Vet International Supplier No. 810877CS
Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Made from recyclable number 5 polypropylene fabric Use for extra heavy instrument sets, loaner trays 4 x 4 feet