Learn More
Med Vet International Surgical Sterilization Pack Wraps, 30" X 30', 50-Pack
Catalog No. 502829741
Encompass_Preferred
These non-woven, soft but tough surgical wraps are useful for wrapping surgery packs during sterilization and storage. They can also be used for actual surgery drapes in precut sizing. The green Sontara fabric is very durable but economical. It withstands steam sterilization for a superior barrier against air and waterborne bacteria.
Catalog No. 50-282-9741 Supplier Med Vet International Supplier No. JOR258C
These non-woven, soft but tough surgical wraps are useful for wrapping surgery packs during sterilization and storage. They can also be used for actual surgery drapes in precut sizing. The green Sontara fabric is very durable but economical. It withstands steam sterilization for a superior barrier against air and waterborne bacteria.