Learn More
Med Vet International Cloth Instrument Wraps, 24"x24", 5/pack
Catalog No. 503042937
Encompass_Preferred
Autoclavable Wraps are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability These two-ply wrappers are stitched with a bias binding that are also color-coded for quick identification
Catalog No. 50-304-2937 Supplier Med Vet International Supplier No. W2424X5
Autoclavable Wraps are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability These two-ply wrappers are stitched with a bias binding that are also color-coded for quick identification