Learn More
Grainger Hallowell Wardrobe Locker, Steel, Louvered, 1 Tier, 15 in. x 21 in. x 78 in., 1 ea
Catalog No. 502793162
Encompass_Preferred
This wardrobe locker is designed for secure storage in professional and industrial environments. Constructed from durable cold rolled steel with a powder coated finish, the single-tier unit features a louvered door design that promotes air circulation to help manage moisture and prevent odor buildup. The locker is supplied fully assembled and includes pre-installed number plates for easy organization.
  • Durable cold rolled steel construction with 24 ga panels and 16 ga door and frame
  • Integrated louvered door design for improved ventilation
  • Recessed lift handle with a padlock hasp for secure closure
  • Includes 6 in. legs for floor clearance
  • Features one shelf and three internal hooks for organized hanging storage
  • Greenguard certified
Catalog No. 50-279-3162 Supplier Grainger Supplier No. 1AEF5
This wardrobe locker is designed for secure storage in professional and industrial environments. Constructed from durable cold rolled steel with a powder coated finish, the single-tier unit features a louvered door design that promotes air circulation to help manage moisture and prevent odor buildup. The locker is supplied fully assembled and includes pre-installed number plates for easy organization.
  • Durable cold rolled steel construction with 24 ga panels and 16 ga door and frame
  • Integrated louvered door design for improved ventilation
  • Recessed lift handle with a padlock hasp for secure closure
  • Includes 6 in. legs for floor clearance
  • Features one shelf and three internal hooks for organized hanging storage
  • Greenguard certified