Learn More
Microsolv Technology Corporation Wrench, 3/8" x 7/16", Open Ended. Flat. 1 EA.
Catalog No. 502271440
Encompass_Preferred
This wrench is chosen for use for HPLC Instruments and Columns due to the size of the flat but also due to the optimal length of the shaft. This length provides sufficient leverage needed for tight fitting without requiring too much force by the user. The opening covers the nuts without excessive over-hang.
This item is not returnable. View return policy
Catalog No. 50-227-1440 Supplier Microsolv Technology Corporation Supplier No. 4603002
This wrench is chosen for use for HPLC Instruments and Columns due to the size of the flat but also due to the optimal length of the shaft. This length provides sufficient leverage needed for tight fitting without requiring too much force by the user. The opening covers the nuts without excessive over-hang.