Learn More
Microsolv Technology Corporation Wrench, 6mm x 8mm, Open Ended. Flare Nut. 1 EA.
Catalog No. 502271595
Encompass_Preferred
This flare nut type wrench is chosen for use for HPLC Instruments that have metric nuts or flats on Columns. This length provides sufficient leverage needed for tight fitting without requiring too much force by the user. This tool is often chosen for use on HPLC Injection Valves.
This item is not returnable. View return policy
Catalog No. 50-227-1595 Supplier Microsolv Technology Corporation Supplier No. 8350180
This flare nut type wrench is chosen for use for HPLC Instruments that have metric nuts or flats on Columns. This length provides sufficient leverage needed for tight fitting without requiring too much force by the user. This tool is often chosen for use on HPLC Injection Valves.