Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wrench, universal, suited for Acros drums, , Thermo Scientific™

Catalog No. AC473470010
Change view
Click to view available options
For Use With (Equipment):
Acros Drums
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
AC473470010 Acros Drums
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC473470010 Supplier Thermo Scientific Chemicals Supplier No. 473470010
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Silver
For Use With (Application) Open Sealed Caps and Drum Seals
For Use With (Equipment) Acros Drums
Material Mild Steel
Dimensions 256 x 100 x 45 mm
Length (Metric) 256 mm
Width (Metric) 100 mm
Height (Metric) 45 mm
Weight (Metric) 1 Kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.