Learn More
Medchemexpress LLC HY-114169 10mg Medchemexpress, WRG-28 CAS:1913291-02-7 Purity:>98%
SDP
Catalog No. 501969297
Encompass_Preferred
Medchemexpress, HY-114169 10mg WRG-28 CAS:1913291-02-7 WRG-28 is a selective, extracellularly acting DDR2 allosteric inhibitor with an IC50 of 230 nM. WRG-28 uniquely inhibits receptor-ligand interactions via allosteric modulation of the receptor. WRG-28 inhibits tumor invasion and migration, as well as tumor-supporting roles of the stroma, and inhibits metastatic breast tumor cell colonization in the lungs by targeting DDR2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9297 Supplier Medchemexpress LLC Supplier No. HY11416910MG
Medchemexpress, HY-114169 10mg WRG-28 CAS:1913291-02-7 WRG-28 is a selective, extracellularly acting DDR2 allosteric inhibitor with an IC50 of 230 nM. WRG-28 uniquely inhibits receptor-ligand interactions via allosteric modulation of the receptor. WRG-28 inhibits tumor invasion and migration, as well as tumor-supporting roles of the stroma, and inhibits metastatic breast tumor cell colonization in the lungs by targeting DDR2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.