Learn More
Volu Sol Wright-Giemsa Stain, 128 oz / 3.78 L, Volu-Sol™
SDP
Supplier:  Volu Sol VWG128
Encompass_Preferred
Romanowsky stain, for peripheral blood and bone marrow smears.  Also used to perform differential white blood cell counts and to study red blood cell morphology. Abnormal granulocyte, lymphocyte or monocyte cell counts may be used to facilitate the diagnosis of diseases such as leukemia or bacterial infections
Catalog No. NC9420039