Learn More
Volu Sol Fuccillo Stain Solution, Wright-Giemsa, 128 oz / 3.78 L, Volu-Sol™
SDP
Catalog No. 501694707 Shop All Volu Sol Products
Encompass_Preferred
Case of 4, Polychromatic stain for peripheral blood and bone marrow smears. Used for red blood cell morphology and white blood cell differentiation. Pre-buffered stain solution.
This item is not returnable. View return policy
Catalog No. 50-169-4707 Supplier Volu Sol Supplier No. VFS128C
Case of 4, Polychromatic stain for peripheral blood and bone marrow smears. Used for red blood cell morphology and white blood cell differentiation. Pre-buffered stain solution.