Learn More
Volu Sol Wright-Giemsa Stain Kit, 8 oz / 250 mL, Volu-Sol™
SDP
Catalog No. NC9494540 Shop All Volu Sol Products
Encompass_Preferred
Wright and Wright-Giemsa Stain Kits include three components:  a stain, buffer, and a rinse.  Stain peripheral blood and  bone marrow smears. Used to perform differential white blood cell counts and to study red blood cell morphology. Abnormal granulocyte, lymphocyte or monocyte cell counts may be used to facilitate the diagnosis of diseases such as leukemia or bacterial infections
Catalog No. NC9494540 Supplier Volu Sol Supplier No. VWG300
Wright and Wright-Giemsa Stain Kits include three components:  a stain, buffer, and a rinse.  Stain peripheral blood and  bone marrow smears. Used to perform differential white blood cell counts and to study red blood cell morphology. Abnormal granulocyte, lymphocyte or monocyte cell counts may be used to facilitate the diagnosis of diseases such as leukemia or bacterial infections