Learn More
StatLab Wright-Giemsa Stain Kit
SDP
Catalog No. STLKTWGIPT
Change view
Click to view available options
Statlab™ Parasite Stain
Catalog No.
STLKTWGIPT Statlab™ Parasite Stain
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. STLKTWGIPT Supplier StatLab Supplier No. KTWGIPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Stains for blood and blood parasites in as little as 2 minutes.
  • Pint stain kit yields between 340-620 slides
  • The MasterTech Optimized Giemsa Stain Kit restores these metachromatic qualities within the dye through the development of a specially formulated Giemsa Buffer, providing exceptional staining results
  • Giemsa serves as the primary stain, producing blue to purple nuclei, cytoplasm, and helicobacter
  • Pint Kit Consists of: Giemsa Stain Stock Pint and Optimized Giemsa Buffer 5 Pints
Specifications
Product Type Stain Kit