Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wright-Giemsa Stain Mixture, Ricca Chemical
SDP

Catalog No. 938016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
938016 500 mL Amber Glass Bottle
938032 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 938016 Supplier Ricca Chemical Company Supplier No. 938016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Wright-Giemsa Stain Mixture
CAS 67-56-1, 56-81-5, 68988-92-1, 51811-82-6, 7558-79-4, 7778-77-0
CAS Min % 83.0, 14.6, 0.32, 0.23, 0.07, 0.04
CAS Max % 86.0, 15.19, 0.33, 0.24, 0.07, 0.04
Packaging Amber Glass Bottle
Color Dark Purple
pH 6.4
Quantity 500 mL
Physical Form Liquid
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.