Learn More
Sigma Aldrich Fine Chemicals Biosciences WRIGHT-GIEMSA STAIN, MOD 500ML
Catalog No. NC2186598
Encompass
Wright-Giemsa Stain, Modified, 0.4% (w/v) in methanol (buffered at pH 6.8) 500mL MFCD00132593
This item is not returnable. View return policy
Catalog No. NC2186598 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. WG16500ML
Wright-Giemsa Stain, Modified, 0.4% (w/v) in methanol (buffered at pH 6.8) 500mL MFCD00132593