Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cenmed™ Wright-Giemsa Stain Pack
SDP

Catalog No. 21520121
Change view
Click to view available options
Quantity:
240 mL Stain
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-121 240 mL Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Flammable Liquid
Catalog No. 21-520-121 Supplier Cenmed™ Supplier No. A371001
Add to Cart
Add to Cart
This item is not returnable. View return policy
Flammable Liquid

Includes

465mL Phosphate Buffer, 240mL Giemsa(5g/L)/Wright Stain(3g/L) Stain, 20% Methyl Alcohol Rinse

Everything you need for your Wright-Giemsa staining procedures in one convienant pack: Stain, Buffer, and Rinse.

Specifications

Quantity 240 mL Stain
Storage Requirements Keep away from open flames. Store away from direct sunlight.
Includes 465mL Phosphate Buffer, 240mL Giemsa(5g/L)/Wright Stain(3g/L) Stain, 20% Methyl Alcohol Rinse
Product Type Stain Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.