Learn More
Description
- Differentiate cell components with the two-part biological stain: Wright-Giesma stain.
- Dark blue to purple liquid with an alcoholic odor.
Specifications
Specifications
| Quantity | 100 mL |
| Storage Requirements | Store in a cool, well-ventilated area away from incompatible substances. Keep away from ignition sources. |
| Format | Liquid |
| For Use With (Application) | Histology |
| Color | Dark Blue to Purple |
| Packaging | HDPE Bottle |
| Assay | Blood |
| CAS | 67-56-1 |
| Molecular Formula | Mixture |
| Synonym | Wright-Giesma Solution |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
