Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wright's Blood Stain, Vion Biosciences
SCI_ED

Catalog No. S02482 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02482 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to Methanol, which is known to the State of California to cause birth defects or other reproductive harm.
Catalog No. S02482 Supplier Aldon Corporation Supplier No. VNWW0060500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to Methanol, which is known to the State of California to cause birth defects or other reproductive harm.

This Wright's blood stain is a hematologic stain to facilitate the differentiation of blood cell types in histological samples.

  • This Wright's blood stain is a hematologic stain to facilitate the differentiation of blood cell types in histological samples.

Specifications

Quantity 500 mL
Description Dark blue liquid with an alcoholic odor.
Storage Requirements Store in a cool, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Boiling Point 65°C (149°F)
Concentration <1%
For Use With (Application) Histology
Color Dark Blue
Melting Point -98°C (-144°F)
Packaging HDPE Bottle
Assay Blood
CAS 67-56-1
Molecular Formula Mixture
Synonym None
Product Type Wright Stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.