Learn More
Medchemexpress LLC Wright's stain | 68988-92-1 | >95.0% | 10 G

Supplier: Medchemexpress LLC HYW08806810G
Wright's stain is a composite hematologic stain used to differentiate blood cell types in peripheral blood and bone marrow smears. It binds nucleic acids and proteins to provide contrast between cellular components for microscopic examination.
- Composite stain that binds nucleic acids and proteins.
- Enables differentiation of erythrocytes, leukocytes, and platelets.
- Suitable for peripheral blood, bone marrow, and cytology smears.
- High purity (≥95.0%) for consistent staining performance.
- Store at 4°C and protect from light; follow SDS/COA for handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.