Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wright's Stain Buffer, Vion Biosciences
SCI_ED

Catalog No. S02480 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02480 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02480 Supplier Aldon Corporation Supplier No. VNWW0070500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Wright's stain buffer yields satisfactory staining results every time when used in tandem with Wright blood stain.

  • This Wright's stain buffer yields satisfactory staining results every time when used in tandem with Wright blood stain.

Specifications

Quantity 500 mL
Description Clear, colorless liquid with a mild characteristic odor.
Storage Requirements Store in a cool, well-ventilated area.
Boiling Point ~100°C (212°F in water)
For Use With (Application) Histology
Color Colorless
Melting Point ~0°C (32°F in water)
pH 6.4
Packaging HDPE
CAS 7732-18-5; 7778-77-0; 7558-79-4
Molecular Formula Mixture
Synonym Giordano pH 6.4
Product Type Buffer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.