Learn More
Electron Microscopy Sciences Wright's Stain, Certified, CI DcWr-38 25 GR
SDP
Catalog No. 5029783
Encompass_Preferred
(2-Propanol)
Solubility: 0.091% Water; 0.23% Alcohol
For differentiation of blood corpuscles. Used with brilliant cresyl blue for staining platelets and reticulum of immature red cells. For staining blood and bone marrow films.
Catalog No. 50-297-83 Supplier Electron Microscopy Sciences Supplier No. 22720
(2-Propanol)
Solubility: 0.091% Water; 0.23% Alcohol
For differentiation of blood corpuscles. Used with brilliant cresyl blue for staining platelets and reticulum of immature red cells. For staining blood and bone marrow films.