Learn More
Medical Chemical Corporation Wright's One Step, 32 oz
SDP
Catalog No. NC1794221
Encompass_Preferred
The traditional stain is diluted 1:1 with giordano buffer before use. Wright's One Step stain contains the buffer already dissolved in the stain. The slides are stained in the undiluted stain and differentiated by decolorizing in purified water.
This item is not returnable. View return policy
Catalog No. NC1794221 Supplier Medical Chemical Corporation Supplier No. 929A32OZ
The traditional stain is diluted 1:1 with giordano buffer before use. Wright's One Step stain contains the buffer already dissolved in the stain. The slides are stained in the undiluted stain and differentiated by decolorizing in purified water.