Learn More
Epredia™ Wright Stain
Catalog No. 22050293 Shop All Epredia Products
Change view
Click to view available options
Quantity:
0.95L
1 gal.
Product Type:
Bottle for Treating Stains
Wright Stain
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-050-293 0.95L Bottle for Treating Stains
22-050-292 1 gal. Wright Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-050-293 Supplier Epredia™ Supplier No. 89012
Add to Cart
Add to Cart
Utilize Epredia™ Wright Stain for peripheral blood and bone marrow smears.
  • Produces results similar to those of the Giemsa stain with a slightly greater contrast
Specifications
Quantity 0.95L
Product Type Bottle for Treating Stains
WARNING: Reproductive Harm - www.P65Warnings.ca.gov