Description
Wright Stain, a modified Romanowsky stain, is intended for differential staining of the cellular elements of blood. It comprises polychrome methylene blue and eosin B. Upon staining blood films with Wright Stain, the white blood cell nucleus and cytoplasm display characteristic blue or pink hues.
Specifications
Specifications
| Quantity | 25 g |
| CAS | 68988-92-1 |
| Product Type | Wright Stain |