Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Wright Stain, Modified

Catalog No. SDWS128
Encompass
Change view
Click to view available options
Quantity:
4L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SDWS128 4L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SDWS128 Supplier Sigma Diagnostics™ Supplier No. WS1284L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for differentially staining the cellular elements of blood

  • Wright stain, 0.3%, buffered at pH 6.8 in methanol
  • Contains stabilizers and surfactant
TRUSTED_SUSTAINABILITY

Specifications

Quantity 4L
Storage Requirements 18-26°C
Format Solution
Concentration 0.3% Buffered in Methanol
For Use With (Application) For dip, rack, and batch staining techniques.
Color Blue
pH 6.75 - 6.95
Product Type Wright Stain, Modified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.