Learn More
Apexbio Technology LLC Wright Staining Solution 500ml
SDP
Catalog No. 5000063453
Encompass_Preferred
Wright Staining Solution composed primarily of eosin and methylene blue is frequently utilized in microscopic examination for differentiation and identification of cell types particularly in hematological analysis Its staining mechanism relies upon the differential affinity of cellular components for acidic and basic dyes eosin an acidic dye stains cytoplasmic granules and erythrocytes pink or orange-red whereas methylene blue a basic dye binds nucleic acids or acidic cellular structures to yield shades of blue or violet In practice mature erythrocytes and eosinophils stain distinctly pink lymphocytes monocytes and basophils appear dark bluish-purple and neutrophils exhibit a pale purple hue due to combined dye affinity Typical usage involves staining thin blood smears or bone marrow aspirates at undiluted formulation to facilitate morphological assessment and differentiation of cell populations
Catalog No. 50-000-63453 Supplier Apexbio Technology LLC Supplier No. K1182500
Wright Staining Solution composed primarily of eosin and methylene blue is frequently utilized in microscopic examination for differentiation and identification of cell types particularly in hematological analysis Its staining mechanism relies upon the differential affinity of cellular components for acidic and basic dyes eosin an acidic dye stains cytoplasmic granules and erythrocytes pink or orange-red whereas methylene blue a basic dye binds nucleic acids or acidic cellular structures to yield shades of blue or violet In practice mature erythrocytes and eosinophils stain distinctly pink lymphocytes monocytes and basophils appear dark bluish-purple and neutrophils exhibit a pale purple hue due to combined dye affinity Typical usage involves staining thin blood smears or bone marrow aspirates at undiluted formulation to facilitate morphological assessment and differentiation of cell populations