Learn More
Andwin Scientific WRIST CRYO GLOVE SM 1 PAIR
SDP
Catalog No. 50930753
Encompass_Preferred
The Cryo-Gloves provide protection from temperature range of 135C to 150C. This product is very comfortable to wear as it is lightweight and breathable. The gloves are ideal for use in ultra low and blast freezers, cold rooms, dry ice handling, liquid nitrogen, liquid oxygen, and liquid helium. This can also be used in several environments, including clinical laboratories and blood banks. This is 12 inches long and is sold in a pair.
This item is not returnable. View return policy
Catalog No. 50-930-753 Supplier Andwin Scientific Supplier No. 71070
The Cryo-Gloves provide protection from temperature range of 135C to 150C. This product is very comfortable to wear as it is lightweight and breathable. The gloves are ideal for use in ultra low and blast freezers, cold rooms, dry ice handling, liquid nitrogen, liquid oxygen, and liquid helium. This can also be used in several environments, including clinical laboratories and blood banks. This is 12 inches long and is sold in a pair.