Learn More
HCL Labels For Research & Development Labs Only - Write-in GHS Label - Kimdura With Matte Lamination - 3.5" x 7" (Pack of 100)
SDP
Catalog No. NC1961788
Encompass_Preferred
For Research & Development (R&D) and Academic Labs Only - Write-in GHS Label - 3.5" x 7" Kimdura With Matte Lamination (Pack of 100) Matte lamination allows for easy writing on the label.These labels are not suitable for production associated laboratories that are required to comply with OSHA's Hazard Communication standard (29 CFR 1910.1200).Perfect for enhancing your Laboratory Safety Program’s labeling system. Complements any “Chemical Hygiene Plan” and “Good Laboratory Safety Practices.” These labels are ideal for identifying chemicals, products, and solutions placed in secondary containers, e.g., beakers, bottles, HPLC solvent containers.IMPORTANT NOTE: These type of labels are for laboratories exempted from complying with OSHA Hazard Communication standard (CFR 29, 1910.1200) but still required to develop and implement a Chemical Hygiene Plan (29CFR 1910.1450). This label design is copyright protected.
Catalog No. NC1961788 Supplier HCL Labels Supplier No. SSL17330003
For Research & Development (R&D) and Academic Labs Only - Write-in GHS Label - 3.5" x 7" Kimdura With Matte Lamination (Pack of 100) Matte lamination allows for easy writing on the label.These labels are not suitable for production associated laboratories that are required to comply with OSHA's Hazard Communication standard (29 CFR 1910.1200).Perfect for enhancing your Laboratory Safety Program’s labeling system. Complements any “Chemical Hygiene Plan” and “Good Laboratory Safety Practices.” These labels are ideal for identifying chemicals, products, and solutions placed in secondary containers, e.g., beakers, bottles, HPLC solvent containers.IMPORTANT NOTE: These type of labels are for laboratories exempted from complying with OSHA Hazard Communication standard (CFR 29, 1910.1200) but still required to develop and implement a Chemical Hygiene Plan (29CFR 1910.1450). This label design is copyright protected.