Learn More
HCL Labels Laboratory Blank Write-In GHS Label - Self-Laminating Adhesive Kimdura - 1.5" x 3.5" (Pack of 250)
SDP
Catalog No. NC1961785
Encompass_Preferred
Laboratory Write-In GHS Label - With Self-Laminating Flap. FOR LABORATORY USE ONLY. This product will enhance your laboratory safety practices as part of your Chemical Hygiene Plan (CHP) or your Best Laboratory Safety Practices implementation programs.Please note that these labels style are not suitable for laboratories that primarily provide quality control analyses for manufacturing processes or that produce hazardous substances for commercial purposes typically referred to as manufacturing or commercial laboratories. Those labs must comply with OSHA's HazCom 2012 (CFR 1910.1200) labeling requirements.
Catalog No. NC1961785 Supplier HCL Labels Supplier No. WIGHS
Laboratory Write-In GHS Label - With Self-Laminating Flap. FOR LABORATORY USE ONLY. This product will enhance your laboratory safety practices as part of your Chemical Hygiene Plan (CHP) or your Best Laboratory Safety Practices implementation programs.Please note that these labels style are not suitable for laboratories that primarily provide quality control analyses for manufacturing processes or that produce hazardous substances for commercial purposes typically referred to as manufacturing or commercial laboratories. Those labs must comply with OSHA's HazCom 2012 (CFR 1910.1200) labeling requirements.