Learn More
Research Products International Corp Write-On Bottles, Wide Mouth, 1000ml, 6 per Package
SDP
Catalog No. 501978394
Encompass
High density polyethylene bottles feature two dedicated surface areas that accept writing from a permanent style marking pen. The defined writing areas provide space for name and date with larger area for detail of bottle contents. Bottle has a wide mouth style for storing liquids or powders. Supplied with leak proof polypropylene cap.
Catalog No. 50-197-8394 Supplier Research Products International Corp Supplier No. 246133
High density polyethylene bottles feature two dedicated surface areas that accept writing from a permanent style marking pen. The defined writing areas provide space for name and date with larger area for detail of bottle contents. Bottle has a wide mouth style for storing liquids or powders. Supplied with leak proof polypropylene cap.