Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Write-On Dry Erase Globe
SCI_ED

Catalog No. S28100 Shop All GSC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric):
32 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
S28100 32 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Not a toy. Product intended for ages 13 and up.

Catalog No. S28100 Supplier GSC Go Science Crazy Supplier No. GLBWO
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Not a toy. Product intended for ages 13 and up.

Helps students learn geography. Globe can be written on with a dry-erase marker, and helps students memorize geographic locations.

  • Made from dry-erase material
  • Set in sturdy plastic base
  • Permanent markings show continents, country/state borders, rivers, lakes, deserts, and currents
  • Latitude and longitude lines shown
  • Marks can easily be erased with a soft cloth

Specifications

Product Type Write-on Erase Globe
Includes 1 Whiteboard globe, 1 plastic base, 1 dry-erase marker
Material Dry-erase Material
Color White
For Use With (Application) Helps learn geography, memorize geographic locations
Subject Geography
Diameter (Metric) 32 cm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.