Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Write-On Inspection Label, Header: Certification, Legend: By, Date, Due

Catalog No. 19112380 Shop All Brady Corporation Products
Change view
Click to view available options
Adhesive Type:
Acrylic, Permanent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Adhesive Type
19-112-380 Acrylic, Permanent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-112-380 Supplier Brady™ Supplier No. 42217
  • $104.50
    $100.65 / Pack of 14

    Save $3.85

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provide easy identification for calibration, testing, and repair of tools and equipment. Brady Write-On Inspection Labels are made of durable, write-on polyester material B-744.

  • Easy identification for calibration, testing, and repair of tools and equipment
  • Made of B-744 write-on polyester
  • Package contains 364 labels

Specifications

Adhesive Type Acrylic, Permanent
Color Black, White
Material Polyester (B-747)
Product Type Write-On Inspection Label
Display Text Legend: CERTIFICATION BY: DATE: DUE:
Shape Rectangle
Size (English) 1.5 x 0.625 in. (W x H)
Size (Metric) 38.1 x 15.8 mm (W x H)
Thickness (English) 0.0034 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.