Learn More
Research Products International Corp Write-On Label Tape Dispenser, Economy
SDP
Catalog No. 501978218
Encompass
Write-On tape dispensers are ideal for organizing and storing assorted rolls of laboratory tape. Hold a variety of tape sizes and colors from 1/2'' to 1'' wide. Accommodates up to five or more rolls of 1/2'' to 3/4'' wide rolls or four 1'' rolls. Removable spindles can be adjusted for 1'' or 3'' cores. Large size platform with serrated cut-off edge provides a convenient writing surface and keeps tape positioned for instant use.
Also features a convenient, built-in marking pen holder and plastic separator discs. The economy model has a plastic cutting edge and mounting holes to secure dispenser in place.
WARNING  This product can expose you to chemicals including Styrene, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-8218 Supplier Research Products International Corp Supplier No. 560171
Write-On tape dispensers are ideal for organizing and storing assorted rolls of laboratory tape. Hold a variety of tape sizes and colors from 1/2'' to 1'' wide. Accommodates up to five or more rolls of 1/2'' to 3/4'' wide rolls or four 1'' rolls. Removable spindles can be adjusted for 1'' or 3'' cores. Large size platform with serrated cut-off edge provides a convenient writing surface and keeps tape positioned for instant use.
Also features a convenient, built-in marking pen holder and plastic separator discs. The economy model has a plastic cutting edge and mounting holes to secure dispenser in place.
WARNING  This product can expose you to chemicals including Styrene, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.