Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Write-On Magnetic Labels

Catalog No. 19112920
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow

Label and update with incredible flexibility. Brady™ Write-On Magnetic Labels can be used for flexible short-term labeling with the included water-soluble pen, or can be used for permanent labeling with a permanent marking pen.

  • Re-useable labels with water-soluble pen (included) that wipes off with wet cloth
  • Special label surface accepts water-soluble pen
  • Magnetic material available in six colors
  • Sold on 1 in. x 50 ft. rolls

Specifications

Color White
Format Roll
Material Magnetic
Product Type Write-On Magnetic Labels
Shape Rectangle
Size (English) 50 ft. x 1 in.
Size (Metric) 15.2 m x 2.54 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.