Learn More
Electron Microscopy Sciences Write Protect Tape, 32 x 13 mm
SDP
Catalog No. 5034210
Encompass_Preferred
Write Protect is a clear adhesive tape designed to cover and protect writing or printing on labels, on the surface of microtubes and on other laboratory containers. The clear tape comes in a handy dispenser and will adhere to all plastics, glass, metals and label materials. The tape is provided in perforated, pull-off segments for 0.5 ml and 1.5 ml tubes. Write Protect will not dry out, peel or fall off and will withstand temperatures ranging from -196 degree C to 150 degree C.
Size: 32 x 13 mm
Catalog No. 50-342-10 Supplier Electron Microscopy Sciences Supplier No. 7762610
Write Protect is a clear adhesive tape designed to cover and protect writing or printing on labels, on the surface of microtubes and on other laboratory containers. The clear tape comes in a handy dispenser and will adhere to all plastics, glass, metals and label materials. The tape is provided in perforated, pull-off segments for 0.5 ml and 1.5 ml tubes. Write Protect will not dry out, peel or fall off and will withstand temperatures ranging from -196 degree C to 150 degree C.
Size: 32 x 13 mm