Learn More
Research Products International Corp Write-On Label Tape, White, 1/2 Inch Roll
SDP
Catalog No. 501367773
Encompass
Write-On label tape adheres to any clean surface including glass, plastics, PTFE, metal and it removes without leaving a sticky residue. Label tape can be marked on with pencil, pen or solvent ink markers. Writing remains legible under the most demanding laboratory conditions. Tape adhesive resists exposure to moisture and most solvents. Withstands temperature extremes from 121°C to -73°C. Rolls have a 3 inch core and contain 120 feet of tape.
Catalog No. 50-136-7773 Supplier Research Products International Corp Supplier No. 560172
Write-On label tape adheres to any clean surface including glass, plastics, PTFE, metal and it removes without leaving a sticky residue. Label tape can be marked on with pencil, pen or solvent ink markers. Writing remains legible under the most demanding laboratory conditions. Tape adhesive resists exposure to moisture and most solvents. Withstands temperature extremes from 121°C to -73°C. Rolls have a 3 inch core and contain 120 feet of tape.