Learn More
Research Products International Corp Write Protect Clear Label for 1.5 - 2.0 ml Tubes, 1.5 x 0.63 Inches, 1000 per Package
SDP
Catalog No. 50212521
Encompass
Write-Protect clear overcoat tape protects markings and labeled surfaces on micro-tubes and test tubes. Tape keeps tube markings and labels intact and prevents smudged, unreadable or lost labels. Adheres to all plastic, glass, metal surfaces and withstands extreme temperature (-196° to +150°C) conditions (i.e. boiling, freezing, thawing, autoclaving, liquid nitrogen). Chemically inert for high resistance to organic solvents, caustic agents, abrasion during vortexing or shaking. Convenient to use. Write on tube or label tube, tear off perforated, pre-cut segment of tape, and place over area. Available in a roll format.
Catalog No. 50-212-521 Supplier Research Products International Corp Supplier No. 247134
Write-Protect clear overcoat tape protects markings and labeled surfaces on micro-tubes and test tubes. Tape keeps tube markings and labels intact and prevents smudged, unreadable or lost labels. Adheres to all plastic, glass, metal surfaces and withstands extreme temperature (-196° to +150°C) conditions (i.e. boiling, freezing, thawing, autoclaving, liquid nitrogen). Chemically inert for high resistance to organic solvents, caustic agents, abrasion during vortexing or shaking. Convenient to use. Write on tube or label tube, tear off perforated, pre-cut segment of tape, and place over area. Available in a roll format.