Learn More
USA Scientific Inc 32 inch Nucleic acid workstation with filtration
Catalog No. NC1707156
Encompass
Combination nucleic acid workstation with HEPA filtration and timer controlled UV light, 32 inW x 24 inD x 30 inH, 110 V
Catalog No. NC1707156 Supplier USA Scientific Inc Supplier No. 42632588
Combination nucleic acid workstation with HEPA filtration and timer controlled UV light, 32 inW x 24 inD x 30 inH, 110 V