Learn More
Aviva Systems Biology Corp WRNIP1 Antibody - N-terminal region (ARP36470_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36470_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WRNIP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WRNIP1. Alternate names: FLJ22526, RP11-420G6.2, WHIP, bA420G6.2ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-014-53048