Learn More
Cayman Chemical WRnr Syndrome rcQ Helcase 25mg
SDP
Catalog No. 502942850
Encompass_Preferred
An inhibitor of WRN (IC50 = 60 nM); selectively inhibits the growth of SW48 cells over DLD-1-WRN-KO cells (GI50s = 70 and >10,000 nM, respectively); reduces tumor volume in an SW48 mouse xenograft model at 240 mg/kg per day
Catalog No. 50-294-2850 Supplier Cayman Chemical Supplier No. 4167925 MG
Add to Cart
Add to Cart
An inhibitor of WRN (IC50 = 60 nM); selectively inhibits the growth of SW48 cells over DLD-1-WRN-KO cells (GI50s = 70 and >10,000 nM, respectively); reduces tumor volume in an SW48 mouse xenograft model at 240 mg/kg per day