Learn More
Medchemexpress LLC HY-P1119 1mg Medchemexpress, WRW4 CAS:878557-55-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP11191MG
Encompass_Preferred
Medchemexpress, HY-P1119 1mg WRW4 CAS:878557-55-2 WRW4, a specific formyl peptide receptor-like 1 (FPRL1) antagonist, inhibits WKYMVm binding to FPRL1 with an IC50 of 0.23 μM. WRW4 specifically inhibits the increase in intracellular calcium by the FPRL1 agonists MMK-1, amyloid beta42 (Abeta42) peptide, and F peptide. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1666